2-Methoxyethyl acrylate ≥98.0% (by GC) stabilised
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Stabilisatie:
Stabilised with 4-methoxyphenol (MEHQ)
A1405-25ML
A1405-500ML
TCIAA1405-25MLEA
18.7
EUR
TCIAA1405-25ML
TCIAA1405-500ML
2-Methoxyethyl acrylate ≥98.0% (by GC) stabilised
2-Methoxyethyl acrylate
Formule:
C₆H₁₀O₃ Kookpunt: 56°C Vlampunt: 60°C |
MDL Nummer:
MFCD00048149 CAS nummer: 3121-61-7 EINECS: 221-499-3 UN: 1992 ADR: 3,III |
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...