1-Acetyl-2-pyrrolidone ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A1418-5G A1418-25G
TCIAA1418-5GEA 58.3 EUR
TCIAA1418-5G TCIAA1418-25G
1-Acetyl-2-pyrrolidone ≥98.0% (by GC)
1-Acetyl-2-pyrrolidone
Formule: C₆H₉NO₂
Kookpunt: 231°C
MDL Nummer: MFCD00014101
CAS nummer: 932-17-2
EINECS: 213-248-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=97338a70df77cad1fd87dbd8f14a34d124f857b8&vid=97338a70df77cad1fd87dbd8f14a34d124f857b8&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Red to Green clear liquid
Purity (GC) min. 98.0 %