Allyltriphenyltin ≥95.0% (W)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

A1491-25G A1491-5G
TCIAA1491-25GEA 235 EUR
TCIAA1491-25G TCIAA1491-5G
Allyltriphenyltin ≥95.0% (W)
Allyltriphenyltin
Formule: C₂₁H₂₀Sn
MDL Nummer: MFCD00048162
CAS nummer: 76-63-1
EINECS: 200-975-4
UN: 3146
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (with ignition residue) min. 95.0 %
Melting point 72.0 to 76.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR