2-Aminobenzotriazole ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

A1585-1G A1585-100MG
TCIAA1585-1GEA 531 EUR
TCIAA1585-1G TCIAA1585-100MG
2-Aminobenzotriazole ≥98.0% (by GC)
2-Aminobenzotriazole
Formule: C₆H₆N₄
MDL Nummer: MFCD00724002
CAS nummer: 1614-11-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 120.0 to 124.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR