3-Amino-4-pyrazolecarbonitrile ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

A1594-25G A1594-5G
TCIAA1594-25GEA 90.2 EUR
TCIAA1594-25G TCIAA1594-5G
3-Amino-4-pyrazolecarbonitrile ≥98.0% (by GC, titration analysis)
3-Amino-4-pyrazolecarbonitrile
Formule: C₄H₄N₄
MDL Nummer: MFCD00005237
CAS nummer: 16617-46-2
EINECS: 240-665-6

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 174.0 to 177.0 deg-C
Solubility in hot Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR