2-Aminophenyl phenyl sulphide ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 2-Aminodiphenyl Sulphide, 2-(Phenylthio)aniline

A1856-25G A1856-500G A1856-100G
TCIAA1856-25GEA 26.4 EUR
TCIAA1856-25G TCIAA1856-500G TCIAA1856-100G
2-Aminophenyl phenyl sulphide ≥98.0% (by GC)
2-Aminophenyl phenyl sulphide
Formule: C₁₂H₁₁NS
MW: 201.29 g/mol
Kookpunt: 258°C
MDL Nummer: MFCD00190139
CAS nummer: 1134-94-7
EINECS: 413-030-8
UN: 3077
ADR: 9 III

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 96.0 %
Melting point 41.0 to 45.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR