4-Amino-5-chloro-2,6-dimethylpyrimidine ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
A1901-25G A1901-5G
TCIAA1901-25GEA 208 EUR
TCIAA1901-25G TCIAA1901-5G
4-Amino-5-chloro-2,6-dimethylpyrimidine ≥98.0% (by GC)
4-Amino-5-chloro-2,6-dimethylpyrimidine
Formule: C₆H₈ClN₃
MDL Nummer: MFCD00191727
CAS nummer: 2858-20-0
Order Now
Specification Test Results
Purity (GC) min. 98.0 %
Melting point 163.0 to 167.0 deg-C