2-Acetyl-1-ethylpyrrole ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

A1977-25G A1977-5G
TCIAA1977-25GEA 71.5 EUR
TCIAA1977-25G TCIAA1977-5G
2-Acetyl-1-ethylpyrrole ≥98.0% (by GC)
2-Acetyl-1-ethylpyrrole
Formule: C₈H₁₁NO
Kookpunt: 82°C
MDL Nummer: MFCD00191234
CAS nummer: 39741-41-8

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR