(3-Acetylphenoxy)acetic acid ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
A2061-25G
A2061-5G
TCIAA2061-25GEA
301
EUR
TCIAA2061-25G
TCIAA2061-5G
(3-Acetylphenoxy)acetic acid ≥98.0% (by GC, titration analysis)
(3-Acetylphenoxy)acetic acid
Formule:
C₁₀H₁₀O₄ |
MDL Nummer:
MFCD02180947 CAS nummer: 1878-80-4 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 117.0 to 121.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...