6-Amyl-2-pyrone ≥96.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
A2147-1G A2147-5G
TCIAA2147-1GEA 70.4 EUR
TCIAA2147-1G TCIAA2147-5G
6-Amyl-2-pyrone ≥96.0% (by GC)
6-Amyl-2-pyrone
Formule: C₁₀H₁₄O₂
Kookpunt: 93°C
Vlampunt: 146°C
MDL Nummer: MFCD00047551
CAS nummer: 27593-23-3
EINECS: 248-552-3
Order Now
Specification Test Results
Appearance Colorless to Brown clear liquid
Purity (GC) min. 96.0 %