Allyl phenoxyacetate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: Phenoxyacetic Acid Allyl Ester

A2177-25G A2177-250G
TCIAA2177-25GEA 33 EUR
TCIAA2177-25G TCIAA2177-250G
Allyl phenoxyacetate ≥98.0% (by GC)
Allyl phenoxyacetate
Formule: C₁₁H₁₂O₃
MW: 192.21 g/mol
Kookpunt: 137°C
MDL Nummer: MFCD00044632
CAS nummer: 7493-74-5
EINECS: 231-335-2
UN: 2810
ADR: 6.1 III

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR