3-Amino-4-hydroxyphenyl methyl sulphone ≥97.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

A2198-25G A2198-5G
TCIAA2198-25GEA 131 EUR
TCIAA2198-25G TCIAA2198-5G
3-Amino-4-hydroxyphenyl methyl sulphone ≥97.0% (by HPLC, titration analysis)
3-Amino-4-hydroxyphenyl methyl sulphone
Formule: C₇H₉NO₃S
MDL Nummer: MFCD00719450
CAS nummer: 98-30-6
EINECS: 202-654-4

Order Now

Specification Test Results

Appearance White to Gray to Brown powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 156.0 to 160.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR