3-Anilino-1-propanol ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: N-(3-Hydroxypropyl)aniline, 3-(Phenylamino)propan-1-ol
A2248-5G A2248-25G
TCIAA2248-5GEA 110 EUR
TCIAA2248-5G TCIAA2248-25G
3-Anilino-1-propanol ≥98.0% (by GC, titration analysis)
3-Anilino-1-propanol
Formule: C₉H₁₃NO
Kookpunt: 140°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD15206074
CAS nummer: 31121-11-6
Order Now
Specification Test Results
Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %