2-Amino-4-chlorobenzonitrile ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

A2483-5G A2483-25G
TCIAA2483-5GEA 46.2 EUR
TCIAA2483-5G TCIAA2483-25G
2-Amino-4-chlorobenzonitrile ≥98.0% (by GC)
2-Amino-4-chlorobenzonitrile
Formule: C₇H₅ClN₂
MDL Nummer: MFCD00035927
CAS nummer: 38487-86-4
EINECS: 253-967-8
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 161.0 to 165.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR