Quinoxalin-6-amine ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
6-Aminoquinoxaline
A2490-1G
A2490-5G
TCIAA2490-1GEA
69.3
EUR
TCIAA2490-1G
TCIAA2490-5G
Quinoxalin-6-amine ≥98.0% (by GC, titration analysis)
Quinoxalin-6-amine
Formule:
C₈H₇N₃ |
MDL Nummer:
MFCD00462821 CAS nummer: 6298-37-9 |
Specification Test Results
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 157.0 to 161.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...