1,4-Anhydro-D-xylitol ≥97.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

A2635-20MG A2635-100MG
TCIAA2635-20MGEA 292 EUR
TCIAA2635-20MG TCIAA2635-100MG
1,4-Anhydro-D-xylitol ≥97.0% (by HPLC)
1,4-Anhydro-D-xylitol
Formule: C₅H₁₀O₄
Kookpunt: 160°C
CAS nummer: 53448-53-6
EINECS: 258-560-9

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange clear liquid
Purity (HPLC) (CAD) min. 97.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR