6-Amino-1-tetralone ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

A2642-1G A2642-5G
TCIAA2642-1GEA 26.4 EUR
TCIAA2642-1G TCIAA2642-5G
6-Amino-1-tetralone ≥98.0% (by GC, titration analysis)
6-Amino-1-tetralone
Formule: C₁₀H₁₁NO
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00099462
CAS nummer: 3470-53-9

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 129.0 to 133.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR