4-Amino-3-(trifluoromethyl)benzonitrile ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
2-Amino-5-cyanobenzotrifluoride
A2644-1G
A2644-5G
TCIAA2644-1GEA
53.9
EUR
TCIAA2644-1G
TCIAA2644-5G
4-Amino-3-(trifluoromethyl)benzonitrile ≥98.0% (by GC)
4-Amino-3-(trifluoromethyl)benzonitrile
Formule:
C₈H₅F₃N₂ |
MDL Nummer:
MFCD00275473 CAS nummer: 327-74-2 UN: 3439 ADR: 6.1,III |
Specification Test Results
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 61.0 to 68.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...