5-Acetyluracil ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

A2670-1G A2670-5G
TCIAA2670-1GEA 45.1 EUR
TCIAA2670-1G TCIAA2670-5G
5-Acetyluracil ≥98.0% (by HPLC, titration analysis)
5-Acetyluracil
Formule: C₆H₆N₂O₃
MDL Nummer: MFCD00151968
CAS nummer: 6214-65-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=215ef9917e74f079c474ef235f6166e04b13d09b&vid=4da022125d2c3c9e48d273d6ec3d8de277032f3f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR