2-(4-Amino-2-nitroanilino)ethanol ≥95.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

A2671-5G A2671-25G
TCIAA2671-5GEA 45.1 EUR
TCIAA2671-5G TCIAA2671-25G
2-(4-Amino-2-nitroanilino)ethanol ≥95.0% (by HPLC, titration analysis)
2-(4-Amino-2-nitroanilino)ethanol
Formule: C₈H₁₁N₃O₃
MDL Nummer: MFCD00071801
CAS nummer: 2871-01-4
EINECS: 220-701-7

Order Now

Specification Test Results

Appearance Dark green to Dark blue to Black powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Nonaqueous Titration) min. 95.0 %
Melting point 124.0 to 128.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR