1,4-Anthraquinone ≥93.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A2686-1G A2686-5G
TCIAA2686-1GEA 61.6 EUR
TCIAA2686-1G TCIAA2686-5G
1,4-Anthraquinone ≥93.0% (by GC)
1,4-Anthraquinone
Formule: C₁₄H₈O₂
Smeltpunt: 218°C
MDL Nummer: MFCD00013724
CAS nummer: 635-12-1
EINECS: 211-228-7
Order Now
Specification Test Results
Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 93.0 %