13-Azido-2,5,8,11-tetraoxatridecane ≥98.0% (by HPLC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A2728-25MG
A2728-100MG
TCIAA2728-25MGEA
36.3
EUR
TCIAA2728-25MG
TCIAA2728-100MG
13-Azido-2,5,8,11-tetraoxatridecane ≥98.0% (by HPLC)
13-Azido-2,5,8,11-tetraoxatridecane
Formule:
C₉H₁₉N₃O₄ Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD13184960 CAS nummer: 606130-90-9 |
|
Specification Test Results
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (HPLC) | min. 98.0 area% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...