4-Amino-2-chlorobenzonitrile ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

A2784-5G A2784-25G
TCIAA2784-5GEA 59.4 EUR
TCIAA2784-5G TCIAA2784-25G
4-Amino-2-chlorobenzonitrile ≥98.0% (by GC)
4-Amino-2-chlorobenzonitrile
Formule: C₇H₅ClN₂
MDL Nummer: MFCD00035926
CAS nummer: 20925-27-3
EINECS: 244-114-0
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 115.0 to 119.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR