2-Acetyl-6-methoxypyridine ≥97.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
A2802-1G A2802-5G
TCIAA2802-1GEA 99 EUR
TCIAA2802-1G TCIAA2802-5G
2-Acetyl-6-methoxypyridine ≥97.0% (by GC)
2-Acetyl-6-methoxypyridine
Formule: C₈H₉NO₂
Kookpunt: 85°C
MDL Nummer: MFCD08276836
CAS nummer: 21190-93-2
Order Now
Specification Test Results
Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 97.0 %
Melting point 39.0 to 43.0 deg-C