3-Acetamidophthalic anhydride ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

A2804-1G A2804-5G
TCIAA2804-1GEA 48.4 EUR
TCIAA2804-1G TCIAA2804-5G
3-Acetamidophthalic anhydride ≥98.0% (by GC, titration analysis)
3-Acetamidophthalic anhydride
Formule: C₁₀H₇NO₄
MDL Nummer: MFCD00453138
CAS nummer: 6296-53-3

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 99.0 %
Purity (Methanolysis Method) min. 98.0 %
Melting point 184.0 to 188.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR