4-Amino-2-chloro-5-fluoropyrimidine ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

A2809-1G A2809-5G
TCIAA2809-1GEA 75.9 EUR
TCIAA2809-1G TCIAA2809-5G
4-Amino-2-chloro-5-fluoropyrimidine ≥98.0% (by GC, titration analysis)
4-Amino-2-chloro-5-fluoropyrimidine
Formule: C₄H₃ClFN₃
MDL Nummer: MFCD00057344
CAS nummer: 155-10-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 193.0 to 197.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR