(R)-(+)-3-Aminoquinuclidine dihydrochloride ≥98.0% (by total nitrogen and titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

A2830-1G A2830-5G
TCIAA2830-1GEA 86.9 EUR
TCIAA2830-1G TCIAA2830-5G
(R)-(+)-3-Aminoquinuclidine dihydrochloride ≥98.0% (by total nitrogen and titration analysis)
(R)-(+)-3-Aminoquinuclidine dihydrochloride
Formule: C₇H₁₆Cl₂N₂
Smeltpunt: 315°C
MDL Nummer: MFCD00191752
CAS nummer: 123536-14-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Specific rotation [a]20/D +22.0 to +26.0 deg (C=1,H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR