5-Amino-3-(4-fluorophenyl)isoxazole ≥95.0% (by HPLC, total nitrogen)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

A2929-200MG A2929-1G
TCIAA2929-200MGEA 83.6 EUR
TCIAA2929-200MG TCIAA2929-1G
5-Amino-3-(4-fluorophenyl)isoxazole ≥95.0% (by HPLC, total nitrogen)
5-Amino-3-(4-fluorophenyl)isoxazole
Formule: C₉H₇FN₂O
MDL Nummer: MFCD03407398
CAS nummer: 81465-82-9

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (with Total Nitrogen) min. 95.0 %
Melting point 100.0 to 104.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR