4-Allylpyrocatechol ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

A2947-10MG A2947-100MG
TCIAA2947-10MGEA 50.6 EUR
TCIAA2947-10MG TCIAA2947-100MG
4-Allylpyrocatechol ≥98.0% (by GC)
4-Allylpyrocatechol
Formule: C₉H₁₀O₂
Kookpunt: 123°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD01548379
CAS nummer: 1126-61-0

Order Now

Specification Test Results

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 43.0 to 49.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR