4-(4-Amino-3-fluorophenoxy)-N-methyl-2-pyridinecarboxamide ≥98.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 4-(4-Amino-3-fluorophenoxy)-N-methylpicolinamide

A2979-200MG A2979-1G
TCIAA2979-200MGEA 26.4 EUR
TCIAA2979-200MG TCIAA2979-1G
4-(4-Amino-3-fluorophenoxy)-N-methyl-2-pyridinecarboxamide ≥98.0% (by HPLC)
4-(4-Amino-3-fluorophenoxy)-N-methyl-2-pyridinecarboxamide
Formule: C₁₃H₁₂FN₃O₂
MW: 261.26 g/mol
MDL Nummer: MFCD09033852
CAS nummer: 757251-39-1

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 95.0 %
Melting point 141.0 to 145.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR