2-Amino-4'-bromoacetophenone hydrochloride ≥98.0% (by HPLC, total nitrogen)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

A3030-1G A3030-5G
TCIAA3030-1GEA 53.9 EUR
TCIAA3030-1G TCIAA3030-5G
2-Amino-4'-bromoacetophenone hydrochloride ≥98.0% (by HPLC, total nitrogen)
2-Amino-4'-bromoacetophenone hydrochloride
Formule: C₈H₉BrClNO
Smeltpunt: 285°C
MDL Nummer: MFCD00051998
CAS nummer: 5467-72-1
EINECS: 226-778-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0db81bc1c069f128e9ebaeeec043566faf4bff41&vid=0db81bc1c069f128e9ebaeeec043566faf4bff41&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR