4-Amino-5,6-dichloropyrimidine

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

A3058-5G A3058-25G
TCIAA3058-5GEA 31.9 EUR
TCIAA3058-5G TCIAA3058-25G
4-Amino-5,6-dichloropyrimidine
4-Amino-5,6-dichloropyrimidine
Formule: C₄H₃Cl₂N₃
MDL Nummer: MFCD09837293
CAS nummer: 310400-38-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d974d2451cd4f2f647f95e666a7f09b49c79712&vid=6d974d2451cd4f2f647f95e666a7f09b49c79712&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR