(4-Aminobenzyl)phosphonic acid ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: (4-Aminobenzyl)phosphonic Acid

A3072-200MG A3072-1G
TCIAA3072-200MGEA 63.8 EUR
TCIAA3072-200MG TCIAA3072-1G
(4-Aminobenzyl)phosphonic acid ≥98.0% (by HPLC, titration analysis)
(4-Aminobenzyl)phosphonic acid
Formule: C₇H₁₀NO₃P
Smeltpunt: 325°C
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00070204
CAS nummer: 5424-27-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=de0bba207a9163e41ff14b730cdb0e3c4386059f&vid=de0bba207a9163e41ff14b730cdb0e3c4386059f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR