(R)-(+)-2-Acetoxypropionyl chloride ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: (R)-1-Chloro-1-oxopropan-2-yl acetate, (+)-O-Acetyl-D-lactoyl chloride

A3085-1G A3085-5G
TCIAA3085-1GEA 85.8 EUR
TCIAA3085-1G TCIAA3085-5G
(R)-(+)-2-Acetoxypropionyl chloride ≥98.0% (by titrimetric analysis)
(R)-(+)-2-Acetoxypropionyl chloride
Formule: C₅H₇ClO₃
Kookpunt: 53°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD27925055
CAS nummer: 53636-19-4
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bb0a4fe3af624fe541985af9f08af32f356ef21a&vid=bb0a4fe3af624fe541985af9f08af32f356ef21a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 96.0 %
Optical purity (GC) min. 96.0 ee%
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR