beta-Alanine Hydroiodide, low water content ≥98.0% (by total nitrogen and titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

A3112-1G A3112-5G
TCIAA3112-1GEA 77 EUR
TCIAA3112-1G TCIAA3112-5G
beta-Alanine Hydroiodide, low water content ≥98.0% (by total nitrogen and titration analysis)
beta-Alanine Hydroiodide
Formule: C₃H₇NO₂·HI
Smeltpunt: 199°C
CAS nummer: 2096495-59-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Water max. 100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR