Alfacalcidol ≥95.0% (by HPLC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
A3125-10MG
A3125-50MG
TCIAA3125-10MGEA
109
EUR
TCIAA3125-10MG
TCIAA3125-50MG
Alfacalcidol ≥95.0% (by HPLC)
Alfacalcidol
Formule:
C₂₇H₄₄O₂ Bewaar temperatuur: Vriezer |
MDL Nummer:
MFCD00868328 CAS nummer: 41294-56-8 EINECS: 255-297-1 UN: 2811 ADR: 6.1,II Merck Index: 14,04819 |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Melting point | 134.0 to 138.0 deg-C |
Specific rotation [a]20/D | +34.0 to +38.0 deg (C=0.3, CHCL3) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...