ABT751 ≥98.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

A3169-10MG A3169-50MG
TCIAA3169-10MGEA 97.9 EUR
TCIAA3169-10MG TCIAA3169-50MG
ABT751 ≥98.0% (by HPLC)
ABT751
Formule: C₁₈H₁₇N₃O₄S
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00910291
CAS nummer: 141430-65-1

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 10.70 to 11.90 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR