2-Acetyl-6-methylpyridine ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

A3214-1G A3214-5G
TCIAA3214-1GEA 47.3 EUR
TCIAA3214-1G TCIAA3214-5G
2-Acetyl-6-methylpyridine ≥98.0% (by GC)
2-Acetyl-6-methylpyridine
Formule: C₈H₉NO
Kookpunt: 83°C
Vlampunt: 84°C
MDL Nummer: MFCD06658292
CAS nummer: 6940-57-4

Order Now

Specification Test Results

Appearance Colorless to Yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR