3-(2-Amino-2-oxoethyl)-5-methylhexanoic acid ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

A3287-25G A3287-100G
TCIAA3287-25GEA 17.6 EUR
TCIAA3287-25G TCIAA3287-100G
3-(2-Amino-2-oxoethyl)-5-methylhexanoic acid ≥98.0% (by HPLC, titration analysis)
3-(2-Amino-2-oxoethyl)-5-methylhexanoic acid
Formule: C₉H₁₇NO₃
MDL Nummer: MFCD08457698
CAS nummer: 181289-15-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 132.0 to 136.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR