Nɑ-(5-Fluoro-2,4-dinitrophenyl)-D-leucinamide ≥98.0% (by HPLC) HPLC labeling reagent for e.e. determination

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

A5524-1G A5524-100MG
TCIAA5524-1GEA 492 EUR
TCIAA5524-1G TCIAA5524-100MG
Nɑ-(5-Fluoro-2,4-dinitrophenyl)-D-leucinamide ≥98.0% (by HPLC) HPLC labeling reagent for e.e. determination
Nalpha-(5-Fluoro-2,4-dinitrophenyl)-D-leucinamide
Formule: C₁₂H₁₅FN₄O₅
MDL Nummer: MFCD03844762
CAS nummer: 178065-30-0
UN: 1325
ADR: 4.1,III

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange powder to crystaline
Purity (HPLC) min. 98.0 area%
Optical purity (LC) min. 99.0 ee%
Melting point 169.0 to 173.0 deg-C
Specific rotation [a]20/D -2.0 to -6.0 deg (C=1, Acetone)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR