4-tert-Butylpyridine ≥96.0% (by GC, titration analysis)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 4-(tert-Butyl)pyridine
B0388-5G B0388-25G
TCIAB0388-5GEA 61.6 EUR
TCIAB0388-5G TCIAB0388-25G
4-tert-Butylpyridine ≥96.0% (by GC, titration analysis)
4-tert-Butylpyridine
Formule: C₉H₁₃N
MW: 135.21 g/mol
Kookpunt: 197°C
Smeltpunt: –40°C
Dichtheid: 0.9219
Vlampunt: 67°C
MDL Nummer: MFCD00006435
CAS nummer: 3978-81-2
EINECS: 223-614-2
Order Now
Specification Test Results
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 96.0 %
Purity (Nonaqueous Titration) min. 96.0 %