Triphenyl borate ≥96.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: Phenyl Borate, Boric Acid Triphenyl Ester

B0524-25G B0524-500G
TCIAB0524-25GEA 88 EUR
TCIAB0524-25G TCIAB0524-500G
Triphenyl borate ≥96.0% (by titrimetric analysis)
Triphenyl borate
Formule: C₁₈H₁₅BO₃
Kookpunt: 158°C
Smeltpunt: 101°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00059011
CAS nummer: 1095-03-0
EINECS: 214-137-0
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) min. 96.0 % (calcd. on exclusive of Boric Acid.)
Boric acid max. 2.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR