Sodium-2-bromoethanesulphonate ≥97.0% (by titrimetric analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
B0589-25G
B0589-500G
TCIAB0589-25GEA
45.1
EUR
TCIAB0589-25G
TCIAB0589-500G
Sodium-2-bromoethanesulphonate ≥97.0% (by titrimetric analysis)
Sodium-2-bromoethanesulphonate
Formule:
C₂H₅BrO₃S |
MDL Nummer:
MFCD00007530 CAS nummer: 4263-52-9 EINECS: 224-244-4 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (Ion exchange titration) | min. 97.0 % |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...