(Bromomethyl)chlorodimethylsilane ≥95.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
BMDMCS, Bromomethyldimethylchlorosilane
B0847-25G
B0847-5G
TCIAB0847-25GEA
453
EUR
TCIAB0847-25G
TCIAB0847-5G
(Bromomethyl)chlorodimethylsilane ≥95.0% (by GC)
(Bromomethyl)chlorodimethylsilane
Formule:
C₃H₈BrClSi Kookpunt: 135°C Vlampunt: 41°C |
MDL Nummer:
MFCD00000503 CAS nummer: 16532-02-8 EINECS: 240-601-7 UN: 2986 ADR: 8,II |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...