N-(Butoxymethyl)acrylamide ≥96.0% (by titrimetric analysis) stabilised

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing


Stabilisatie: Stabilised with 4-methoxyphenol (MEHQ)

B1060-25ML B1060-500ML
TCIAB1060-25MLEA 17.6 EUR
TCIAB1060-25ML TCIAB1060-500ML
N-(Butoxymethyl)acrylamide ≥96.0% (by titrimetric analysis) stabilised
N-(Butoxymethyl)acrylamide
Formule: C₈H₁₅NO₂
MW: 157.21 g/mol
Vlampunt: 78°C
MDL Nummer: MFCD00059417
CAS nummer: 1852-16-0
EINECS: 217-442-7

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (Bromination Method) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR