Lawesson's reagent ≥95.0% (by titrimetric analysis) sulphurating reagent
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
2,4-Bis(4-methoxyphenyl)-2,4-dithioxo-1,3,2,4- dithiadiphosphetane, 2,4-Bis(4-methoxyphenyl)-1,3-dithia-2,4-diphosphetane 2,4-Disulphide
B1133-25G
B1133-250G
TCIAB1133-25GEA
27.5
EUR
TCIAB1133-25G
TCIAB1133-250G
Lawesson's reagent ≥95.0% (by titrimetric analysis) sulphurating reagent
Lawesson's reagent
Formule:
C₁₄H₁₄O₂P₂S₄ |
MDL Nummer:
MFCD00005171 CAS nummer: 19172-47-5 EINECS: 242-855-4 Merck Index: 14,05391 |
Specification Test Results
Appearance | White to Light yellow to Green powder to crystal |
Purity (Iodometric Titration) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...