1-Bromo-2-methoxymethylnaphthalene ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: 1-Bromo-2-(methoxymethyl)naphthalene

B1170-5G B1170-25G
TCIAB1170-5GEA 109 EUR
TCIAB1170-5G TCIAB1170-25G
1-Bromo-2-methoxymethylnaphthalene ≥98.0% (by GC)
1-Bromo-2-methoxymethylnaphthalene
Formule: C₁₂H₁₁BrO
Kookpunt: 151°C
MDL Nummer: MFCD00059518
CAS nummer: 64689-70-9

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR