Bis(2,2,2-trifluoroethyl)ether ≥99.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

B1293-1G B1293-5G
TCIAB1293-1GEA 34.1 EUR
TCIAB1293-1G TCIAB1293-5G
Bis(2,2,2-trifluoroethyl)ether ≥99.0% (by GC)
Bis(2,2,2-trifluoroethyl)ether
Formule: C₄H₄F₆O
Kookpunt: 64°C
Vlampunt: 1°C
MDL Nummer: MFCD00010604
CAS nummer: 333-36-8
UN: 3271
ADR: 3,II
Merck Index: 14,04201

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR