N-tert-Butyl-2-methoxyethylamine ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
N-(2-Methoxyethyl)-tert-butylamine
B1430-1G
B1430-5G
TCIAB1430-1GEA
45.1
EUR
TCIAB1430-1G
TCIAB1430-5G
N-tert-Butyl-2-methoxyethylamine ≥98.0% (by GC)
N-tert-Butyl-2-methoxyethylamine
Formule:
C₇H₁₇NO MW: 131.22 g/mol Kookpunt: 137°C |
MDL Nummer:
MFCD00145008 UN: 2733 ADR: 3 III |
|
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...