3-Bromobutyronitrile ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

B1452-25G B1452-5G
TCIAB1452-25GEA 190 EUR
TCIAB1452-25G TCIAB1452-5G
3-Bromobutyronitrile ≥98.0% (by GC)
3-Bromobutyronitrile
Formule: C₄H₆BrN
Kookpunt: 72°C
MDL Nummer: MFCD00039488
CAS nummer: 20965-20-2
UN: 3276
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR